Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ampicillin (trihydrate) | 7177-48-2 | 99.9% | 5 G
SDP

Catalog No. 5000407958
Encompass_Preferred

Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic effective against gram-positive and gram-negative bacteria. It inhibits E. coli growth, alleviates hemorrhagic enteritis symptoms, and provides neuroprotection against ischemia-reperfusion brain injury by reducing MMPs activities and increasing GLT-1 expression.

  • Molecular weight: 403.45
  • Molecular formula: C16H25N3O7S
  • Appearance: White to off-white solid

Catalog No. 50-004-07958 Supplier Medchemexpress LLC Supplier No. HYB0522B5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic effective against gram-positive and gram-negative bacteria. It inhibits E. coli growth, alleviates hemorrhagic enteritis symptoms, and provides neuroprotection against ischemia-reperfusion brain injury by reducing MMPs activities and increasing GLT-1 expression.

  • Molecular weight: 403.45
  • Molecular formula: C16H25N3O7S
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.