Learn More
Medchemexpress LLC Ampicillin trihydrate | 7177-48-2 | 99.9% | 50 G
SDP
Catalog No. 5000405555
Encompass_Preferred
Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria. It is intended for research use only.
  • Inhibits the growth of E. coli of swine origin in a dose-dependent manner at 2.5 μg/mL.
  • Effective in alleviating the symptoms of hemorrhagic enteritis in 11-week-old pigs.
  • Produces maximum concentrations in bile twice as high as in serum.
  • Provides neuroprotection against ischemia-reperfusion brain injury by reducing MMPs activities and increasing GLT-1 expression.
Catalog No. 50-004-05555 Supplier Medchemexpress LLC Supplier No. HYB0522B50G
Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria. It is intended for research use only.
  • Inhibits the growth of E. coli of swine origin in a dose-dependent manner at 2.5 μg/mL.
  • Effective in alleviating the symptoms of hemorrhagic enteritis in 11-week-old pigs.
  • Produces maximum concentrations in bile twice as high as in serum.
  • Provides neuroprotection against ischemia-reperfusion brain injury by reducing MMPs activities and increasing GLT-1 expression.