Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ampicillin trihydrate | 7177-48-2 | MFCD00072036 | 99.9% | 500 MG
SDP

Supplier:  Medchemexpress LLC HYB0522B500MG

Encompass_Preferred

Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic effective against various gram-positive and gram-negative bacteria. It is supplied as a white to off-white solid with a purity of 99.92%.

  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive bacteria
  • Effective against gram-negative bacteria
  • High purity of 99.92%
  • Available in multiple pack sizes
  • Stable when stored at 4°C, sealed, away from moisture and light

Catalog No. 50-004-08567


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.