Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampicillin Trihydrate, MP Biomedicals™

Catalog No. MP219014725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219014725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219014725 Supplier MP Biomedicals, Inc Supplier No. 0219014725
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Active as a broad-spectrum antibiotic. Ampicillin Trihydrate, MP Biomedicals is a semi-synthetic derivative of penicillin that is commonly used to select for ampicillin resistance in mutated and transformed cells. It is also used for enhancing luminol che

Ampicillin trihydrate is a β-lactam antibiotic with an amino group side chain attached to the penicillin structure. It is a penicillin derivative that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane.

  • Inactivated by beta-lactamases; therefore, a beta-lactamase inhibitor should be considered when using ampicillin
  • Antimicrobial spectrum: gram-negative and gram-positive bacteria.
    • Against gram-positive bacteria, ampicillin has a similar mode of action as benzylpenicillin
    • Against gram-negative bacteria, it has a similar mode of action as chloramphenicol and tetracyclines
  • Bactericidal only to growing E. coli. Inhibits cell wall synthesis via cleavage of β-lactam ring of ampicillin by β-lactamase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7177-48-2
Molecular Formula C16H25N3O7S
Molecular Weight (g/mol) 403.45
InChI Key RXDALBZNGVATNY-CWLIKTDRSA-N
Synonym Aminobenzylpenicillin trihydrate, alpha-Aminobenzylpenicillin trihydrate ;D[-]-α-Aminobenzylpenicillin, 6-[D(-)-α-Aminophenylacetamido]penicillanic acid
PubChem CID 23565
ChEBI CHEBI:31209
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.O.O.O

Specifications

Physical Form Powder
Melting Point 198°C to 200°C (literature)
Color White
Concentration Working Concentration: 100mg/liter for both gram positive and gram negative bacteria. It is typically stable in media at 37°C for approximately 3 days.
Quantity 25 g
Solubility Information Soluble in 1M Ammonium Hydroxide (50mg/mL), dilute acids (50mg/mL 1M HCl) or bases; slightly soluble in water (10mg/mL); practically insoluble in alcohol, chloroform, ether and fixed oils.
Formula Weight 403.46
Grade Not Available
Recommended Storage Store at +4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.