Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampicillin Trihydrate, USP Grade, G-Biosciences
SCI_ED

Catalog No. S139816
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139816 100 g
S162505 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139816 Supplier G-Biosciences Supplier No. RC887
Add to Cart
Add to Cart

Ampicillin is a member of the extended-spectrum Beta-lactam family and similar in structure to penicillin. It is used in the selection of antibiotic cell hybrids and has a high rate of action against many gram-negative bacterial strains.

Ampicillin sodium targets non-extended spectrum Beta-lactamase bacteria including Staphylococcus and Streptococcus species and medically important enteric pathogens such as Shigella and Salmonella. Resistance to ampicillin is routinely utilized as a selectable marker to confirm successful cell transformation.

Chemical Identifiers

CAS 7177-48-2

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 100 g
Percent Purity 98%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.