Learn More
AdipoGen Ampicilline sodium salt
SDP
Catalog No. 502561750
Encompass_Preferred
Chemical. CAS 69-52-3. Formula C16H18N3NaO4S. MW 371.39. Ampicillin is a broad-spectrum antibiotic with activity against Gram-positive and Gram-negative bacteria. Formulations containing ampicillin have been used to treat a variety of bacterial infections. This is a beta-lactam antibiotic that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Administration with beta-lactamase cleaves the beta-lactam ring of Ampicillin and inactivates it. Ampicillin is widely used in cell culture as a selective agent for transformed cells expressing beta-lactamase. Can also be used as reference material.
Catalog No. 50-256-1750 Supplier AdipoGen Supplier No. CDXA0313G005
Chemical. CAS 69-52-3. Formula C16H18N3NaO4S. MW 371.39. Ampicillin is a broad-spectrum antibiotic with activity against Gram-positive and Gram-negative bacteria. Formulations containing ampicillin have been used to treat a variety of bacterial infections. This is a beta-lactam antibiotic that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Administration with beta-lactamase cleaves the beta-lactam ring of Ampicillin and inactivates it. Ampicillin is widely used in cell culture as a selective agent for transformed cells expressing beta-lactamase. Can also be used as reference material.