Learn More
Crescent Chemical Co Inc AMPICILLINTRIHYDRATE RES.GRADE
Supplier: Crescent Chemical Co Inc SE13397.01
Ampicillin trihydrate research grade, Ph. Eur. Semi-synthetic derivative of penicillin. Inhibitor of cell wall synthesis in E. coli. Soluble 1 part in 150 parts H2O as well as in diluted acids and bases. Insoluble in alcohol. Size - 10G Storage Conditions - +1