Learn More
Crescent Chemical Co Inc AMPICILLINTRIHYDRATE RES.GRADE
SDP
Catalog No. 50247292
Encompass_Preferred
Ampicillin trihydrate research grade, Ph. Eur. Semi-synthetic derivative of penicillin. Inhibitor of cell wall synthesis in E. coli. Soluble 1 part in 150 parts H2O as well as in diluted acids and bases. Insoluble in alcohol. Size - 100G Storage Conditions - +
Catalog No. 50-247-292 Supplier Crescent Chemical Co Inc Supplier No. SE13397.03
Ampicillin trihydrate research grade, Ph. Eur. Semi-synthetic derivative of penicillin. Inhibitor of cell wall synthesis in E. coli. Soluble 1 part in 150 parts H2O as well as in diluted acids and bases. Insoluble in alcohol. Size - 100G Storage Conditions - +