Learn More
Eurofins Discoverx AMPK (α2, β1, γ1), active, 250 µg. (14-902M)
Supplier:  Eurofins Discoverx 14902M
Encompass
Recombinant, human AMPK (α2, β1, γ1) holoenzyme, comprised of full length N-terminal 6xHis-tagged AMPKα2, full length untagged AMPKβ1 and full length untagged AMPKγ1. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9390