Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AMPK (A1/B1/G3), His Tag Recombinant Protein

Catalog No. PV6241
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6241 10 μg
PV6243 1 mg
PV6242 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV6241 Supplier Invitrogen™ Supplier No. PV6241
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human AMPK (combination of A1/B1/G3 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for the three subunits (A1/B1/G3) are NP_006242.5, NP_006244.2, and NP_059127.2.

AMPK (A2/B2/G3) is a member of the AMPK family which are heterotrimeric proteins consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPKs are an important energy-sensing enzyme group in the cells that monitor energy status particularly in response to stress. AMPKs regulate fatty acid and cholesterol synthesis by regulating the key rate-limiting enzymes acetyl-CoA carboxylase and hydroxy beta-methylglutaryl-CoA reductase. The beta subunit may be a positive regulator of AMPK activity and is highly expressed in skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13131, Q9UGI9, Q9Y478
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM sodium phosphate with 0.2mM DTT, 25% glycerol, 150mM Imidazole, 300mM NaCl, 0.1mM PMSF and no preservative; pH 7
Gene ID (Entrez) 53632, 5562, 5564
Molecular Weight (g/mol) 63.6-38-51 kDa
Name Human AMPK (A1/B1/G3), His Tag
pH Range 7
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AMPK alpha 1/beta 1/gamma 3; AMPK alpha1/beta1/gamma3
Common Name AMPK alpha-1/beta-1/gamma-3
Gene Symbol PRKAA1, PRKAB1, PRKAG3
Protein Length aa 10-end
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence 10-end
Content And Storage Storage Buffer: 50 mM sodium phosphate pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol

Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Assay Entry Biochemical binding/kinetic
Kinase Group Serine/Threonine Kinase
Target Entry AMPK (A1/B1/G3)
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Fragment
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Drug Discovery: Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.