Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AMPK A2/B1/G1, GST Tag, His Tag Recombinant Protein

Catalog No. PV4674
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4674 10 μg
PV4675 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV4674 Supplier Invitrogen™ Supplier No. PV4674
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

AMP-activated protein kinase (AMPK) is a heterotrimeric complex that acts as sensor of cellular energy levels. The signaling cascades initiated by activating AMPK are critical to regulating metabolic events in the liver, skeletal muscle, heart, adipose tissue, and pancreas. An impairment in fuel metabolism that occurs in obesity is a factor leading to type 2 diabetes. The insulin resistance associated with type 2 diabetes is most profound at the level of skeletal muscle, the primary site of glucose and fatty acid utilization. Activation of AMPK is of interest for the treatment of type 2 diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P54619, P54646, Q9Y478
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 2mM DTT, 50% glycerol, 0.5mM EDTA, 150mM NaCl, 0.02% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 5563, 5564, 5571
Molecular Weight (g/mol) 91.4-57.8-41.7 kDa
Name Human AMPK A2/B1/G1, GST Tag, His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AMPK alpha 2/beta 1/gamma 1; AMPK alpha2/beta1/gamma1
Common Name AMPK alpha-2/beta-1/gamma-1
Gene Symbol PRKAA2, PRKAB1, PRKAG1
Kinase Family CAMKL Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag, His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: CAMK Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Tag Position GST (N-Terminal)⁄GST (N-Terminal)⁄His (N-Terminal)
Target Entry AMPK (A2/B1/G1)
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.