Learn More
Invitrogen™ Human AMPK alpha-2/beta-2/gamma-1, His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human AMPK (combination of A2/B2/G1 subunits) was expressed by baculovirus in Sf9 insect cells using C-terminal His tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant
Specifications
Specifications
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM sodium phosphate with 150mM Imidazole, 0.2mM DTT, 25% glycerol, 300mM NaCl, 0.1mM PMSF and no preservative; pH 7 |
| Molecular Weight (g/mol) | 63.1-36-40 kDa |
| Name | Human AMPK alpha-2/beta-2/gamma-1, His Tag |
| pH Range | 7 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Storage Requirements | -80°C, Avoid Freeze/Thaw Cycles |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.