Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AMPK alpha-2/beta-2/gamma-2, His Tag Recombinant Protein

Catalog No. PV6250
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6250 10 μg
PV6251 100 μg
PV6252 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV6250 Supplier Invitrogen™ Supplier No. PV6250
Add to Cart
Edge
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human AMPK (combination of A2/B2/G2 subunits) was expressed by baculovirus in Sf9 insect cells using C-terminal His tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant

AMPK (A2/B2/G2) is a member of the AMPK family which are heterotrimeric proteins consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPKs are an important energy-sensing enzyme group in the cells that monitor energy status particularly in response to stress. AMPKs regulate fatty acid and cholesterol synthesis by regulating the key rate-limiting enzymes acetyl-CoA carboxylase and hydroxy beta-methylglutaryl-CoA reductase. The beta subunit may be a positive regulator of AMPK activity and is highly expressed in skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM sodium phosphate with 0.1 mM PMSF, 0.2 mM DTT, 150 mM Imidazole, 25% glycerol, 300 mM NaCl and no preservative; pH 7
Molecular Weight (g/mol) 63.1-36-41 kDa
Name Human AMPK alpha-2/beta-2/gamma-2, His Tag
Quantity 10 μg
Common Name AMPK alpha-2/beta-2/gamma-2
Protein Length 1-552, 1-272, 1-569
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.