Learn More
Sino Biological Recombinant Human AMPK Protein (full length, His Tag)
Catalog No. 5000083646
Encompass_Preferred
Recombinant full-length human AMPK (combination of A2/B2/G3 subunits) was expressed by baculovirus in Sf9 insect cells using C-terminal His tags.
Catalog No. 50-000-83646 Supplier Sino Biological Supplier No. P4610H50
Recombinant full-length human AMPK (combination of A2/B2/G3 subunits) was expressed by baculovirus in Sf9 insect cells using C-terminal His tags.