Learn More
Sino Biological Recombinant Human AMPK (G1/B1/A1) Heterotrimer Protein (Full Length, A1/B1/G1, His Tag), Active
Catalog No. 502573658
Encompass_Preferred
Recombinant full length human AMPK (combination of A1/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using C-terminal His tags.
Catalog No. 50-257-3658 Supplier Sino Biological Supplier No. P4710H500
Recombinant full length human AMPK (combination of A1/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using C-terminal His tags.