Learn More
Sino Biological Recombinant Human AMPK (A2/B2/G3) Heterotrimer Protein (Full Length, A2/B2/G3, His & GST Tag)
Catalog No. 502573661
Encompass_Preferred
Recombinant full length human AMPK (combination of A2/B2/G3 subunits) was expressed by baculovirus in Sf9 insect cells using the N-terminal GST and C-terminal His tags.
Catalog No. 50-257-3661 Supplier Sino Biological Supplier No. P4614GH100
Recombinant full length human AMPK (combination of A2/B2/G3 subunits) was expressed by baculovirus in Sf9 insect cells using the N-terminal GST and C-terminal His tags.