Learn More
Medchemexpress LLC AMPK activator 12 | 431920-24-0 | 99.68% | 426.35 | 5 MG
SDP
Catalog No. 5000359339
Encompass_Preferred
AMPK activator 12 is a potent AMP-activated protein kinase (AMPK) activator and GDF15 inducer. It increases GDF15 protein levels in human hepatic cells.
  • Potent AMPK activator
  • GDF15 inducer
  • Increases GDF15 protein levels in human hepatic cells
Catalog No. 50-003-59339 Supplier Medchemexpress LLC Supplier No. HY1494065MG
AMPK activator 12 is a potent AMP-activated protein kinase (AMPK) activator and GDF15 inducer. It increases GDF15 protein levels in human hepatic cells.
  • Potent AMPK activator
  • GDF15 inducer
  • Increases GDF15 protein levels in human hepatic cells