Learn More
Medchemexpress LLC AMPK activator 12 | 431920-24-0 | 99.7% | 426.35 | 1 ML
SDP
Catalog No. 5000361050
Encompass_Preferred
AMPK activator 12 (compound 21) is a potent AMPK activator and GDF15 inducer. It increases GDF15 protein levels in human hepatic cells.
  • Potent AMPK activator
  • GDF15 inducer
  • Increases GDF15 protein levels
  • Suitable for research use
Catalog No. 50-003-61050 Supplier Medchemexpress LLC Supplier No. HY14940610MM/1ML
AMPK activator 12 (compound 21) is a potent AMPK activator and GDF15 inducer. It increases GDF15 protein levels in human hepatic cells.
  • Potent AMPK activator
  • GDF15 inducer
  • Increases GDF15 protein levels
  • Suitable for research use