Learn More
TARGETMOL CHEMICALS INC AMPK activator 12 5MG
SDP
Catalog No. 502879735
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AMPK activator 12 is an AMPK activator and GDF15 inducer that elevates the expression level of GDF15 protein in human hepatocytes, which can be used for cancer research. Purity 99.27%
Catalog No. 50-287-9735 Supplier TARGETMOL CHEMICALS INC Supplier No. T787915MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AMPK activator 12 is an AMPK activator and GDF15 inducer that elevates the expression level of GDF15 protein in human hepatocytes, which can be used for cancer research. Purity 99.27%