Learn More
Medchemexpress LLC AMPK activator 2 hydrochloride | 00-00-0 | 99.7% | 337.77 | C13H19ClF3N5 | 5 MG
SDP
Catalog No. 5000263963
Encompass_Preferred
AMPK activator 2 hydrochloride is a fluorine-containing proguanil derivative that activates AMP-activated protein kinase (AMPK), modulating downstream mTOR/4EBP1/p70S6K signaling. It is reported to inhibit proliferation and migration of human cancer cell lines and is provided as a research reagent for in vitro studies.
  • Up-regulates AMPK signaling pathway.
  • Downregulates mTOR/4EBP1/p70S6K signaling.
  • Inhibits proliferation and migration in UMUC3, T24, and A549 cell lines.
  • Available in small quantities suitable for assay development.
  • High reported purity for biochemical experiments.
Catalog No. 50-002-63963 Supplier Medchemexpress LLC Supplier No. HY130723A5MG
AMPK activator 2 hydrochloride is a fluorine-containing proguanil derivative that activates AMP-activated protein kinase (AMPK), modulating downstream mTOR/4EBP1/p70S6K signaling. It is reported to inhibit proliferation and migration of human cancer cell lines and is provided as a research reagent for in vitro studies.
  • Up-regulates AMPK signaling pathway.
  • Downregulates mTOR/4EBP1/p70S6K signaling.
  • Inhibits proliferation and migration in UMUC3, T24, and A549 cell lines.
  • Available in small quantities suitable for assay development.
  • High reported purity for biochemical experiments.