Learn More
Medchemexpress LLC AMPK activator 2 hydrochloride | 00-00-0 | 99.7% | C13H19ClF3N5 | 10MG
SDP
Catalog No. 5000260632
Encompass_Preferred
AMPK activator 2 hydrochloride is a small-molecule research reagent that activates the AMPK signaling pathway and suppresses mTOR/4EBP1/p70S6K signaling. It has demonstrated inhibition of proliferation and migration in human cancer cell lines (UMUC3, T24, A549) and is supplied as a white to off-white solid for in vitro biochemical and cellular studies.
  • Activates AMPK signaling for pathway modulation studies.
  • Downregulates mTOR/4EBP1/p70S6K to assess downstream effects.
  • Demonstrated antiproliferative and antimigratory activity in cancer cell lines.
  • High purity suitable for biochemical and cellular assays.
  • Solid, stable formulation with defined storage recommendations.
Catalog No. 50-002-60632 Supplier Medchemexpress LLC Supplier No. HY130723A10MG
AMPK activator 2 hydrochloride is a small-molecule research reagent that activates the AMPK signaling pathway and suppresses mTOR/4EBP1/p70S6K signaling. It has demonstrated inhibition of proliferation and migration in human cancer cell lines (UMUC3, T24, A549) and is supplied as a white to off-white solid for in vitro biochemical and cellular studies.
  • Activates AMPK signaling for pathway modulation studies.
  • Downregulates mTOR/4EBP1/p70S6K to assess downstream effects.
  • Demonstrated antiproliferative and antimigratory activity in cancer cell lines.
  • High purity suitable for biochemical and cellular assays.
  • Solid, stable formulation with defined storage recommendations.