Learn More
TARGETMOL CHEMICALS INC AMPK ACTIVATOR 4 5MG
SDP
Catalog No. 502579480
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMPK activator 4 is a potent and selective AMPK activator that does not inhibit mitochondrial complex I. AMPK activator 4 selectively activates AMPK in muscle tissue and dose-dependently improves glucose tolerance in normal mice significantly reduces fasting glucose levels and improves insulin resistance in db/db diabetic mice.AMPK activator 4 has hypoglycemic effects. activator 4 has hypoglycemic effects. purity: 99%
Catalog No. 50-257-9480 Supplier TARGETMOL CHEMICALS INC Supplier No. T622385MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMPK activator 4 is a potent and selective AMPK activator that does not inhibit mitochondrial complex I. AMPK activator 4 selectively activates AMPK in muscle tissue and dose-dependently improves glucose tolerance in normal mice significantly reduces fasting glucose levels and improves insulin resistance in db/db diabetic mice.AMPK activator 4 has hypoglycemic effects. activator 4 has hypoglycemic effects. purity: 99%