Learn More
Medchemexpress LLC HY-131334 5mg Medchemexpress, AMPK activator 4 CAS:2493239-46-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1313345MG
Encompass_Preferred
Medchemexpress, HY-131334 5mg AMPK activator 4 CAS:2493239-46-4 AMPK activator 4 is a potent AMPK activator without inhibition of mitochondrial complex I. AMPK activator 4 selectively activates AMPK in the muscle tissues. AMPK activator 4 dose-dependently improves glucose tolerance in normal mice, and significantly lowers fasting blood glucose level and ameliorates insulin resistance in db/db diabetic mice. Anti-hyperglycemic effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1272