Learn More
Cayman Chemical AMPK actIvatr 10mg
SDP
Catalog No. 502970168
Encompass_Preferred
An indirect activator of AMPK (EC50 = 11.7 μM); stimulates glucose uptake in L6 myocytes and promotes phosphorylation of ACC, a known target of AMPK; decreases blood glucose concentrations in Zucker diabetic rats, demonstrating its utility in whole animals; inhibits the growth of multiple myeloma cells
Catalog No. 50-297-0168 Supplier Cayman Chemical Supplier No. 1474110 MG
An indirect activator of AMPK (EC50 = 11.7 μM); stimulates glucose uptake in L6 myocytes and promotes phosphorylation of ACC, a known target of AMPK; decreases blood glucose concentrations in Zucker diabetic rats, demonstrating its utility in whole animals; inhibits the growth of multiple myeloma cells