Learn More
Cayman Chemical AMPK activator, Purity: ≥95%, CAS: 849727-81-7, 5 mg
SDP
Catalog No. 502970408
Encompass_Preferred
An indirect activator of AMPK (EC50 = 11.7 μM); stimulates glucose uptake in L6 myocytes and promotes phosphorylation of ACC, a known target of AMPK; decreases blood glucose concentrations in Zucker diabetic rats, demonstrating its utility in whole animals; inhibits the growth of multiple myeloma cells
Catalog No. 50-297-0408 Supplier Cayman Chemical Supplier No. 147415 MG
An indirect activator of AMPK (EC50 = 11.7 μM); stimulates glucose uptake in L6 myocytes and promotes phosphorylation of ACC, a known target of AMPK; decreases blood glucose concentrations in Zucker diabetic rats, demonstrating its utility in whole animals; inhibits the growth of multiple myeloma cells