Learn More
Sigma Aldrich Fine Chemicals Biosciences AMPK alpha1 beta1 gamma1 P
Catalog No. 502906072
Encompass
Recombinant human AMPK (alpha1 beta1 gamma1) holoenzyme comprised of full length N-terminal 6His-tagged AMPKalpha1 full length untagged AMPKbeta1 and full length untagged AMPKgamma1.
Catalog No. 50-290-6072 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 14840
Add to Cart
Add to Cart
Recombinant human AMPK (alpha1 beta1 gamma1) holoenzyme comprised of full length N-terminal 6His-tagged AMPKalpha1 full length untagged AMPKbeta1 and full length untagged AMPKgamma1.