Learn More
Cell Signaling Technology AMPK and ACC Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502729464
Encompass
The AMPK and ACC Antibody Sampler Kit provides an economical means to investigate energy homeostasis and fatty acid synthesis within the cell. This kit includes a comprehensive selection of primary and secondary antibodies designed to facilitate the study of AMP-activated protein kinase (AMPK) signaling and Acetyl-CoA carboxylase (ACC) function. Each component is optimized to perform two Western blots.
  • Includes primary and secondary antibodies for AMPK and ACC targets
  • Validated for Western blotting applications
  • Suitable for investigations of energy homeostasis and fatty acid synthesis pathways
  • Supplied in a ready-to-use format for laboratory research
Catalog No. 50-272-9464 Supplier Cell Signaling Technology Supplier No. 9957T
The AMPK and ACC Antibody Sampler Kit provides an economical means to investigate energy homeostasis and fatty acid synthesis within the cell. This kit includes a comprehensive selection of primary and secondary antibodies designed to facilitate the study of AMP-activated protein kinase (AMPK) signaling and Acetyl-CoA carboxylase (ACC) function. Each component is optimized to perform two Western blots.
  • Includes primary and secondary antibodies for AMPK and ACC targets
  • Validated for Western blotting applications
  • Suitable for investigations of energy homeostasis and fatty acid synthesis pathways
  • Supplied in a ready-to-use format for laboratory research