Learn More
Cayman Chemical AMPK Complexa11nd 1 10ug
SDP
Catalog No. 502950967
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged AMPK complex (α1, β1, and γ1 subunits) expressed in insect cells • Amino acids: 1-559, 1-270, and 1-331 for the α1, β1, and γ1 subunits, respectively • MW: ~146 kDa
Catalog No. 50-295-0967 Supplier Cayman Chemical Supplier No. 3255710 UG
Source: Active recombinant human C-terminal His-tagged AMPK complex (α1, β1, and γ1 subunits) expressed in insect cells • Amino acids: 1-559, 1-270, and 1-331 for the α1, β1, and γ1 subunits, respectively • MW: ~146 kDa