Learn More
Medchemexpress LLC AMPK gamma 1 Antibody (YA621) | affinity purified | 38 kDa | 50 UL
SDP
Catalog No. 5000395873
Encompass_Preferred
AMPK gamma 1 antibody (YA621) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting AMPK gamma 1. It is supplied in PBS, pH 7.4, and is suitable for various applications including WB, ICC/IF, and IP. This antibody is designed for research use only and has not been fully validated for medical applications.
  • Reacts with human samples
  • Monoclonal antibody from rabbit host
  • Non-conjugated
  • Suitable for Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunoprecipitation (IP)
  • Stored at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles
  • Appears as a liquid
Catalog No. 50-003-95873 Supplier Medchemexpress LLC Supplier No. HYP8054350UL
AMPK gamma 1 antibody (YA621) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting AMPK gamma 1. It is supplied in PBS, pH 7.4, and is suitable for various applications including WB, ICC/IF, and IP. This antibody is designed for research use only and has not been fully validated for medical applications.
  • Reacts with human samples
  • Monoclonal antibody from rabbit host
  • Non-conjugated
  • Suitable for Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunoprecipitation (IP)
  • Stored at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles
  • Appears as a liquid