Learn More
Medchemexpress LLC Ampk gamma 1/beta 1/alpha 1 heterotrimer protein, human (sf9, His-GST) | >=98.0% | 160 kDa | 100 UG
SDP
Catalog No. 5000399060
Encompass_Preferred
The AMPK gamma 1/beta 1/alpha 1 Heterotrimer Protein, Human is a recombinant human-derived protein, expressed by Sf9 insect cells with N-His, N-GST labeled tag. As a crucial regulator of cellular energy metabolism, it is activated by low ATP levels, which enhances energy production while inhibiting biosynthetic processes and cell growth. The heterotrimer serves as the AMP/ATP-binding subunit of AMPK, influencing activation or inhibition through its gamma non-catalytic subunit.
  • Recombinant human-derived protein
  • Expressed by Sf9 insect cells
  • N-His, N-GST labeled tag
  • Regulator of cellular energy metabolism
  • Activated by low ATP levels
  • Enhances energy production
  • Inhibits biosynthetic processes and cell growth
  • Mediates binding to AMP, ADP, and ATP
  • Modulates cellular polarity by remodeling the actin cytoskeleton
  • Heterotrimeric composition includes alpha, beta, and gamma subunits
Catalog No. 50-003-99060 Supplier Medchemexpress LLC Supplier No. HYP76141100UG
The AMPK gamma 1/beta 1/alpha 1 Heterotrimer Protein, Human is a recombinant human-derived protein, expressed by Sf9 insect cells with N-His, N-GST labeled tag. As a crucial regulator of cellular energy metabolism, it is activated by low ATP levels, which enhances energy production while inhibiting biosynthetic processes and cell growth. The heterotrimer serves as the AMP/ATP-binding subunit of AMPK, influencing activation or inhibition through its gamma non-catalytic subunit.
  • Recombinant human-derived protein
  • Expressed by Sf9 insect cells
  • N-His, N-GST labeled tag
  • Regulator of cellular energy metabolism
  • Activated by low ATP levels
  • Enhances energy production
  • Inhibits biosynthetic processes and cell growth
  • Mediates binding to AMP, ADP, and ATP
  • Modulates cellular polarity by remodeling the actin cytoskeleton
  • Heterotrimeric composition includes alpha, beta, and gamma subunits