Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AMPK gamma 1/beta 1/alpha 1 Heterotrimer Protein, Human (sf9, His-GST) | ≥98.0% | Approximately 38 kDa (PRKAB1) & 40 kDa (PRKAG1) & 95 kDa (PRKAA1), based on SDS-PAGE under reducing conditions. | 5 UG
SDP

Catalog No. 5000400199
Encompass_Preferred

AMPK gamma 1/beta 1/alpha 1 Heterotrimer Protein, Human (sf9, His-GST) is a recombinant human-derived protein expressed by Sf9 insect cells with N-His and N-GST labeled tags. It functions as the AMP/ATP-binding subunit of AMP-activated protein kinase (AMPK), a critical regulator of cellular energy metabolism. Activated by low ATP levels, AMPK enhances energy production while inhibiting biosynthetic processes and cell growth, thereby maintaining cellular energy homeostasis.

  • Specific activity determined to be 53 nmol/min/mg
  • Expressed in Sf9 insect cells
  • N-His and N-GST labeled tags
  • Purity greater than 98.0% by SDS-PAGE
  • Endotoxin level less than 1 EU/μg
  • Stable at -80°C for 1 year from receipt
  • Formulated as a 0.22 μm filtered solution of 20 mM Tris, 300 mM NaCl, 10% glycerol, pH 7.5
  • Approximate molecular weights: 38 kDa (PRKAB1), 40 kDa (PRKAG1), 95 kDa (PRKAA1)

Catalog No. 50-004-00199 Supplier Medchemexpress LLC Supplier No. HYP761415UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMPK gamma 1/beta 1/alpha 1 Heterotrimer Protein, Human (sf9, His-GST) is a recombinant human-derived protein expressed by Sf9 insect cells with N-His and N-GST labeled tags. It functions as the AMP/ATP-binding subunit of AMP-activated protein kinase (AMPK), a critical regulator of cellular energy metabolism. Activated by low ATP levels, AMPK enhances energy production while inhibiting biosynthetic processes and cell growth, thereby maintaining cellular energy homeostasis.

  • Specific activity determined to be 53 nmol/min/mg
  • Expressed in Sf9 insect cells
  • N-His and N-GST labeled tags
  • Purity greater than 98.0% by SDS-PAGE
  • Endotoxin level less than 1 EU/μg
  • Stable at -80°C for 1 year from receipt
  • Formulated as a 0.22 μm filtered solution of 20 mM Tris, 300 mM NaCl, 10% glycerol, pH 7.5
  • Approximate molecular weights: 38 kDa (PRKAB1), 40 kDa (PRKAG1), 95 kDa (PRKAA1)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.