Learn More
Medchemexpress LLC AMPK gamma 1/beta 2/alpha 1 heterotrimer protein | 96.3% | 50 UG
SDP
Catalog No. 5000293993
Encompass_Preferred
Recombinant human AMPK gamma 1/beta 2/alpha 1 heterotrimer produced in Sf9 insect cells with an N-terminal His-GST tag. Supplied as a purified protein preparation for in vitro biochemical and structural studies, it enables activity assays, binding experiments, and protein-protein interaction analyses. Typical reported purity is 96.3%, and the product is provided as a 50 μg vial.
  • Human origin for physiologically relevant studies.
  • Produced in Sf9 insect cells for proper folding and complex assembly.
  • N-terminal His-GST tag for affinity purification and detection.
  • High purity (96.3%) suitable for biochemical assays.
  • Provided as a 50 μg aliquot for small-scale experiments.
  • Approximate subunit molecular weights: ~35 kDa, ~37 kDa, ~95 kDa.
Catalog No. 50-002-93993 Supplier Medchemexpress LLC Supplier No. HYP7614350UG
Recombinant human AMPK gamma 1/beta 2/alpha 1 heterotrimer produced in Sf9 insect cells with an N-terminal His-GST tag. Supplied as a purified protein preparation for in vitro biochemical and structural studies, it enables activity assays, binding experiments, and protein-protein interaction analyses. Typical reported purity is 96.3%, and the product is provided as a 50 μg vial.
  • Human origin for physiologically relevant studies.
  • Produced in Sf9 insect cells for proper folding and complex assembly.
  • N-terminal His-GST tag for affinity purification and detection.
  • High purity (96.3%) suitable for biochemical assays.
  • Provided as a 50 μg aliquot for small-scale experiments.
  • Approximate subunit molecular weights: ~35 kDa, ~37 kDa, ~95 kDa.