Learn More
Medchemexpress LLC AMPK gamma 1/beta 2/alpha 1 heterotrimer protein, human | 96.3% | 10 UG
SDP
Catalog No. 5000293860
Encompass_Preferred
Recombinant human AMPK α1β2γ1 heterotrimer protein expressed in Sf9 insect cells with an N-terminal His-GST tag. Supplied as a purified recombinant complex for in vitro kinase assays, biochemical characterization, and screening of modulators that affect AMPK activity.
  • Expressed in Sf9 insect cells with an N-terminal His-GST tag.
  • Purity approximately 96.3%.
  • Subunit molecular weights approx. 95 kDa (α1), 37 kDa (β2), 35 kDa (γ1).
  • Validated for in vitro kinase assays and biochemical profiling.
  • Supplied in microgram quantities suitable for biochemical assays.
Catalog No. 50-002-93860 Supplier Medchemexpress LLC Supplier No. HYP7614310UG
Recombinant human AMPK α1β2γ1 heterotrimer protein expressed in Sf9 insect cells with an N-terminal His-GST tag. Supplied as a purified recombinant complex for in vitro kinase assays, biochemical characterization, and screening of modulators that affect AMPK activity.
  • Expressed in Sf9 insect cells with an N-terminal His-GST tag.
  • Purity approximately 96.3%.
  • Subunit molecular weights approx. 95 kDa (α1), 37 kDa (β2), 35 kDa (γ1).
  • Validated for in vitro kinase assays and biochemical profiling.
  • Supplied in microgram quantities suitable for biochemical assays.