Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AMPK gamma 1/beta 2/alpha 2 heterotrimer protein | >90.0% | 10 UG
SDP

Catalog No. 5000293996
Encompass_Preferred

Recombinant human AMPK gamma 1/beta 2/alpha 2 heterotrimer protein produced in Sf9 insect cells with N-terminal His and GST tags. Supplied as a 0.22 μm filtered solution in Tris-NaCl buffer with stabilizers, the protein shows high purity and measurable kinase activity suitable for biochemical assays and structural studies. Store frozen and avoid repeated freeze-thaw cycles.

  • Recombinant heterotrimeric AMPK composed of gamma 1, beta 2, and alpha 2 subunits.
  • Expressed in Sf9 insect cells for eukaryotic folding and post-translational processing.
  • N-terminal His and GST tags for affinity purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Supplied in a stabilized buffer formulation containing Tris, NaCl, EDTA, DTT, glycerol, and antioxidants.
  • Specific activity of 1-6 nmol/min/mg measured using SAMS peptide.
  • Available in small aliquots to minimize freeze-thaw and preserve activity.

Catalog No. 50-002-93996 Supplier Medchemexpress LLC Supplier No. HYP7614410UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human AMPK gamma 1/beta 2/alpha 2 heterotrimer protein produced in Sf9 insect cells with N-terminal His and GST tags. Supplied as a 0.22 μm filtered solution in Tris-NaCl buffer with stabilizers, the protein shows high purity and measurable kinase activity suitable for biochemical assays and structural studies. Store frozen and avoid repeated freeze-thaw cycles.

  • Recombinant heterotrimeric AMPK composed of gamma 1, beta 2, and alpha 2 subunits.
  • Expressed in Sf9 insect cells for eukaryotic folding and post-translational processing.
  • N-terminal His and GST tags for affinity purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Supplied in a stabilized buffer formulation containing Tris, NaCl, EDTA, DTT, glycerol, and antioxidants.
  • Specific activity of 1-6 nmol/min/mg measured using SAMS peptide.
  • Available in small aliquots to minimize freeze-thaw and preserve activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.