Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ampk gamma 1/beta 2/alpha 2 heterotrimer protein (human) | >90.0% | 100 UG
SDP

Catalog No. 5000293954
Encompass_Preferred

Recombinant human AMPK heterotrimer containing α2, β2, and γ1 subunits, expressed in Sf9 insect cells and supplied as a His- and GST-tagged protein solution for research applications. The preparation is supplied at research-grade purity and formulated in a stabilized Tris/NaCl buffer with reducing agents and glycerol for improved stability during storage and handling.

  • Expression system: Sf9 insect cells.
  • Fusion tags: N-terminal His and GST.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in 50 mM Tris, 200 mM NaCl, 1 mM EDTA, 1 mM DTT, 0.5 mM PMSF, 10% glycerol, 1 mM GSH, pH 7.4.
  • Store at -80 °C; avoid repeated freeze-thaw cycles.
  • For research use only.

Catalog No. 50-002-93954 Supplier Medchemexpress LLC Supplier No. HYP76144100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human AMPK heterotrimer containing α2, β2, and γ1 subunits, expressed in Sf9 insect cells and supplied as a His- and GST-tagged protein solution for research applications. The preparation is supplied at research-grade purity and formulated in a stabilized Tris/NaCl buffer with reducing agents and glycerol for improved stability during storage and handling.

  • Expression system: Sf9 insect cells.
  • Fusion tags: N-terminal His and GST.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in 50 mM Tris, 200 mM NaCl, 1 mM EDTA, 1 mM DTT, 0.5 mM PMSF, 10% glycerol, 1 mM GSH, pH 7.4.
  • Store at -80 °C; avoid repeated freeze-thaw cycles.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.