Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AMPK gamma 1/beta 2/alpha 2 heterotrimer protein, human (Sf9, His-GST) | >90.0% | 50 UG
SDP

Catalog No. 5000293917
Encompass_Preferred

Recombinant human AMPK heterotrimer (α2, β2, γ1) expressed in Sf9 insect cells, supplied as a filtered solution with N-terminal His and GST tags for biochemical and structural studies. The preparation is formulated in a stabilizing buffer and provided for research use only.

  • Source organism: human.
  • Expression system: Sf9 insect cells.
  • Tags: N-terminal His and GST.
  • Purity: >90.0% by reducing SDS-PAGE.
  • Formulation: filtered solution in 50 mM Tris, 200 mM NaCl, 1 mM EDTA, 1 mM DTT, 10% glycerol, pH 7.4.
  • Storage: store at -80 °C for long-term and avoid repeated freeze-thaw cycles.
  • Intended use: for research use only.

Catalog No. 50-002-93917 Supplier Medchemexpress LLC Supplier No. HYP7614450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human AMPK heterotrimer (α2, β2, γ1) expressed in Sf9 insect cells, supplied as a filtered solution with N-terminal His and GST tags for biochemical and structural studies. The preparation is formulated in a stabilizing buffer and provided for research use only.

  • Source organism: human.
  • Expression system: Sf9 insect cells.
  • Tags: N-terminal His and GST.
  • Purity: >90.0% by reducing SDS-PAGE.
  • Formulation: filtered solution in 50 mM Tris, 200 mM NaCl, 1 mM EDTA, 1 mM DTT, 10% glycerol, pH 7.4.
  • Storage: store at -80 °C for long-term and avoid repeated freeze-thaw cycles.
  • Intended use: for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.