Learn More
Cayman Chemical AMPKa1 RabbIt MoncLNal ntI
SDP
Catalog No. 502949194
Encompass_Preferred
A mixture of ganglioside GD3 with variable fatty acyl chains induces apoptosis in HuT-78 cutaneous T cell lymphoma cells in a concentration-dependent manner and disrupts the mitochondrial membrane potential at 200 M expression in GD3-negative SK-MEL-28-N1 malignant melanoma cells increases both cell proliferation and invasion in vitro ganglioside GD3-deficient adult mice exhibit progressive loss of the NSC pool and impaired neurogenesis
Catalog No. 50-294-9194 Supplier Cayman Chemical Supplier No. 32246100 UL
A mixture of ganglioside GD3 with variable fatty acyl chains induces apoptosis in HuT-78 cutaneous T cell lymphoma cells in a concentration-dependent manner and disrupts the mitochondrial membrane potential at 200 M expression in GD3-negative SK-MEL-28-N1 malignant melanoma cells increases both cell proliferation and invasion in vitro ganglioside GD3-deficient adult mice exhibit progressive loss of the NSC pool and impaired neurogenesis