Learn More
LGC GENOMICS LLC (Lucigen) Ampligase 10X Reaction Buffer, 5 mL
Catalog No. NC9808447
Encompass
Ampligase Thermostable DNA Ligase catalyzes NAD-dependent ligation of adjacent 3-hydroxylated and 5-phosphorylated termini in duplex DNA structures that are stable at high temperatures
This item is not returnable. View return policy
Catalog No. NC9808447 Supplier LGC GENOMICS LLC Supplier No. A1905B
Ampligase Thermostable DNA Ligase catalyzes NAD-dependent ligation of adjacent 3-hydroxylated and 5-phosphorylated termini in duplex DNA structures that are stable at high temperatures