Learn More
Amplifex™ Keto Reagent Kit , MilliporeSigma™ Supelco™
Catalog No. 111004090 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
:
1 Kit
Catalog No.
11-100-4090 1 Kit
1 options
Catalog No. 11-100-4090 Supplier MilliporeSigma Supelco Supplier No. 44659621KT
Only null left
Add to Cart
Add to Cart
Kit contains a protocol, 2 tubes of Amplifex™ Keto reagent and 2 tubes of diluent. Each tube contains enough reagent to complete 25 reactions.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Amplifex is a trademark of AB Sciex Pte. Ltd.