Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Amplify Setting Solution
SDP

Catalog No. 22445526 Shop All Mopec Products
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-445-526 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-445-526 Supplier Mopec™ Supplier No. BG100
Add to Cart
Add to Cart

Amplify is a fixative spray usually applied after the application of Harmony dyes, although it can also be applied to tissue prior to marking with dyes. The fixative spray modifies the surface of challenging specimens to aid the adhesion of Harmony dyes.

  • Strongly recommended to use Ink Fix for use with fatty specimens
  • Can be applied to tissue before its marked with dye
  • Suitable for fatty specimens

Specifications

Capacity 250 mL
For Use With (Equipment) Tissue Marking Dyes
Grade Chemical Solution
Product Type Fixative Spray
Quantity 1 Bottle
Solution Type Fixative
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.