Learn More
LGC GENOMICS LLC (Lucigen) Ampligase DNA Ligase without Buffer, 10,000 U @ 100 U/LHighly thermostable DNA ligase for applications where ligation at high temperature is beneficial or required.
Catalog No. NC0923742
Encompass
Ampligase Thermostable DNA Ligase catalyzes NAD-dependent ligation of adjacent 3-hydroxylated and 5-phosphorylated termini in duplex DNA structures that are stable at high temperaturesHigh thermostability allows ligation using high-stringency hybridization conditionsHigh specificity and stringency permits sensitive detection of SNPsAvailable in kit and "enzyme only" formats11
This item is not returnable. View return policy
Catalog No. NC0923742 Supplier LGC GENOMICS LLC Supplier No. A0110K
Ampligase Thermostable DNA Ligase catalyzes NAD-dependent ligation of adjacent 3-hydroxylated and 5-phosphorylated termini in duplex DNA structures that are stable at high temperaturesHigh thermostability allows ligation using high-stringency hybridization conditionsHigh specificity and stringency permits sensitive detection of SNPsAvailable in kit and "enzyme only" formats11