Learn More
LGC GENOMICS LLC (Lucigen) Ampligase DNA Ligase with Buffer, 2,500 U @ 5 U/LHighly thermostable DNA ligase for applications where ligation at high temperature is beneficial or required.
Catalog No. NC1516788
Encompass
Ampligase Thermostable DNA Ligase catalyzes NAD-dependent ligation of adjacent 3-hydroxylated and 5-phosphorylated termini in duplex DNA structures that are stable at high temperatures
Catalog No. NC1516788 Supplier LGC GENOMICS LLC Supplier No. A3202K
Ampligase Thermostable DNA Ligase catalyzes NAD-dependent ligation of adjacent 3-hydroxylated and 5-phosphorylated termini in duplex DNA structures that are stable at high temperatures