Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ AMPLIPUR™ Expert Tips

Catalog No. F174200G Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 20 μL
10 to 200 μL
100 to 1000 μL
100 to 1200 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
F174200G 1 to 20 μL
F174100G 0.5 to 10 μL
F174300G 10 to 200 μL
F174400G 100 to 1000 μL
F174500G 100 to 1200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. F174200G Supplier Gilson™ Supplier No. F174200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compatible with most pipettes and compliant with ISO 8655, they are made from recyclable materials and offer durable, high-performance pipetting at excellent value.

  • Sterility and Cleanliness for Reliable Results: Ensure the integrity of your experiments and the reliability of your data. AmpliPur™ Expert Tips, sterile version, are certified with a Sterility Assurance Level (SAL) of 10−6 and are free from Human DNA/RNA and DNase/RNase. Ideal for microbiology, molecular biology, biomedicine, cell culture, and biobanking, they provide the high level of cleanliness and sterility required for your most critical applications.
  • Sustainable Design for a Greener Future: AmpliPur™ Expert Tips and their packaging are made from 94% recyclable materials, supporting your commitment to sustainability while reducing environmental impact.Their eco-friendly design ensures that your work not only meets the highest scientific standards but also contributes to a more sustainable future.
  • Excellent Fit With Added Convenience: AmpliPur™ Expert Tips, sterile version, are designed to fit most common pipettes found in the laboratory (ISO 8655 compliant).The AmpliPur Expert Tips can address the needs of advanced biological workflows.

Specifications

Volume (Metric) 1 to 20 μL
Filtered Filtered
Sterility Sterile
Color Yellow
Autoclavable Not Autoclavable
Format Rack, 10 x 96
Tip Style Standard
Product Type Pipette Tips
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line AmpliPur
Material Polyethylene
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.