Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ AmpliPur™ Expert Tips PROMO

Catalog No. F174201GPM Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
10 to 20 μL
10 to 200 μL
100 to 1000 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
F174201GPM 10 to 20 μL
F174301GPM 10 to 200 μL
F174401GPM 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F174201GPM Supplier Gilson™ Supplier No. F174201PM

AmpliPur™ Expert Tips are ISO 8655 compliant and provide a significant value for the money spent. These filter tips protect precious samples and your pipettes from cross-contamination and are designed to fit various pipette brands.

  • AmpliPur Expert Tips are designed to fit various pipette brands for maximum convenience on the bench.
  • Equipped with a filter, they also help prevent cross-contamination.
  • Ideal for Molecular Biology.
  • These tips significantly contribute to molecular testing performance by ensuring sample integrity during the pipetting process.
  • Manufactured according to the certified Gilson quality standard, AmpliPur Expert Tips comply with ISO 8655 requirements and are certified free of RNase and DNase, preserving your sample integrity.

Specifications

Volume (Metric) 10 to 20 μL
Filtered Filtered
Sterility Non-sterile
Color Clear
Rack Color Yellow
Autoclavable Not Autoclavable
Length (English) 2 in.
Length (Metric) 52 mm
Certifications/Compliance ISO 8655
Clean Claims Yes
For Use With (Application) Molecular Biology
Format Rack
Tip Style Specialty
Packaging Tipack
Quantity 960/Pk.
Product Type Pipette Tips
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Product Line AmpliPur
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.