Learn More
LGC GENOMICS LLC AmpliScribe T7 High Yield Kit, High Yield In Vitro Transcription, RNase Inhibitor Included, 50 Reactions
Catalog No. NC2162035
Encompass
The AmpliScribe™ T7 High Yield Transcription Kit by Biosearch Technologies enables efficient in vitro transcription for high yields of RNA transcripts. It uniquely supports the synthesis of both long and short RNA molecules using high concentrations of NTPs, producing over 20-fold more full-length RNA compared to conventional systems. With scalability to milligram amounts and labeling capabilities for fluorescent, biotinylated, or digoxigenin RNA, it is ideal for versatile RNA applications.
This item is not returnable. View return policy
Catalog No. NC2162035 Supplier LGC GENOMICS LLC Supplier No. AS3107
The AmpliScribe™ T7 High Yield Transcription Kit by Biosearch Technologies enables efficient in vitro transcription for high yields of RNA transcripts. It uniquely supports the synthesis of both long and short RNA molecules using high concentrations of NTPs, producing over 20-fold more full-length RNA compared to conventional systems. With scalability to milligram amounts and labeling capabilities for fluorescent, biotinylated, or digoxigenin RNA, it is ideal for versatile RNA applications.