Learn More
Bio Rad Laboratories AMPLISIZE MOLECULAR RULER
Catalog No. 509012883
Encompass
Criterion Tris-HCl gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 13.3 cm x 8.7 cm x 1.0 mm (W x L x thickness) are supplied in a 15.0 x 10.6 cm cassette and are compatible with Criterio.
Catalog No. 50-901-2883 Supplier Bio Rad Laboratories Supplier No. 1708200
Criterion Tris-HCl gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 13.3 cm x 8.7 cm x 1.0 mm (W x L x thickness) are supplied in a 15.0 x 10.6 cm cassette and are compatible with Criterio.