Learn More
Aat Bioquest Amplite Colorimetric Superoxide Dismutase SOD Assay Kit (200 Tests)
SDP
Supplier:  Aat Bioquest 11305
Encompass_Preferred
Superoxide dismutases SOD are a class of enzymes that catalyze the dismutation of superoxide into oxygen and hydrogen peroxide. This catalog is for 200 Tests.
Catalog No. 50-224-9421