Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec AMPLITUDE LAMBDA WIPE

Catalog No. 17103016 Shop All Contec Inc. Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
17-103-016 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-103-016 Supplier Contec Supplier No. AMLA0013
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amplitude™ Lambda™ wipes are made from a hydroentangled blend of 58% cellulose and 42% polyester. Ideal for general cleaning in biotech, pharma, semiconductor, and electronics industries.

  • Hydroentangled cellulose/polyester
  • Excellent strength and durability
  • Provides low levels of particles and extractables
  • Excellent for wiping and absorbing spills

Specifications

Product Type Dry Wipe
Length (English) 7 in.
Length (Metric) 17.78 cm
Width (English) 7 in.
Width (Metric) 17.78 cm
Material Cellulose
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.