Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec Amplitude Pi Microfiber Wipe

Catalog No. 17720590 Shop All Contec Inc. Products
Change view
Click to view available options
Material:
Microfiber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17-720-590 Microfiber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-720-590 Supplier Contec Supplier No. AMPI0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amplitude Pi wipes are designed to help life science manufacturers control contamination and ultimately maintain cGMP compliance while consuming less resources.

  • Nonwoven split microfiber increases cleaning effectiveness and contaminant pick up and offers a smooth, even laydown of solvent to surfaces
  • Covers greater surface area compared to non-microfiber wipes, reducing the number of wipes required for some cleaning tasks. The wipes are made from polyester and nylon split microfiber, enhancing their cleaning effectiveness and contaminant pick-up ability.

Specifications

Product Type Microfiber Wipe
Length (English) 9 in.
Length (Metric) 230 mm
Width (English) 9 in.
Width (Metric) 230 mm
Material Microfiber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.