Learn More
Contec™ Amplitude™ Standard Weight Heatseal Knit Wipes
BIOPHARMAGREENER_CHOICE
Catalog No. 19033455 Shop All Contec Inc. Products
Change view
Click to view available options
Packaging Quantity:
150 Ea./Bag, 10 Bags/Cs. (Bulk)
300/Bag, 5 Bags/Cs. (Bulk)
75 Wipes/Bag, 16 Bags/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19-033-455 150 Ea./Bag, 10 Bags/Cs. (Bulk)
19-130-5950 300/Bag, 5 Bags/Cs. (Bulk)
19-038-735 75 Wipes/Bag, 16 Bags/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-033-455 Supplier Contec™ Supplier No. AMVL0004
Add to Cart
Add to Cart
This item is not returnable. View return policy
Clean surfaces effectively without scratching. Contec™ Amplitude™ Standard Weight Heatseal Knit Wipes are made with fabric knitted from filament polyester for a soft, textured surface.
  • Soft, textured wipe surface for effective cleaning without scratching
  • Laser cut, which seals the fibers at edges
  • Made with fabric knitted from filament polyester
  • Packaged in an ISO Class 4 cleanroom to further reduce contamination
Specifications
Product Type Heatseal Wipe
Dimensions (L x W) 9 x 9 in. (22.86 x 22.86 cm)
Packaging Quantity 150 Ea./Bag, 10 Bags/Cs. (Bulk)